Ethyl-4-bromo-3-oxobutanoate 90%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 4-bromoacetoacetate, Ethyl 4-bromo-3-oxobutanoate
OR4299-1G
OR4299-25G
OR4299-5G
APOSOR4299-1GEA
36.4
EUR
APOSOR4299-1G
APOSOR4299-25G
APOSOR4299-5G
Ethyl-4-bromo-3-oxobutanoate 90%
Ethyl-4-bromo-3-oxobutanoate
Formula:
C₆H₉BrO₃ MW: 209,0379 g/mol Boiling Pt: 70…71 °C/1mmHg Density: 1,528 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD07783015 CAS Number: 13176-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...