4-Bromo-3-ethoxyaniline hydrochloride 98%

Supplier: Apollo Scientific
Warning

OR4344-25G OR4344-100G
APOSOR4344-25GEA 145 EUR
APOSOR4344-25G APOSOR4344-100G
4-Bromo-3-ethoxyaniline hydrochloride 98%
4-Bromo-3-ethoxyaniline hydrochloride
Formula: C₈H₁₁BrClNO
MW: 216,08 g/mol
Melting Pt: 186…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03790891
CAS Number: 846023-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR