2-(5-Amino-1H-indazol-1-yl)ethanol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Amino-1-(2-hydroxyethyl)-1H-indazole

OR43551-1G OR43551-250MG
APOSOR43551-1GEA 435 EUR
APOSOR43551-1G APOSOR43551-250MG
2-(5-Amino-1H-indazol-1-yl)ethanol 97%
2-(5-Amino-1H-indazol-1-yl)ethanol
Formula: C₉H₁₁N₃O
Storage Temperature: Ambient
CAS Number: 885270-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR