6-Amino-1-(tetrahydropyranyl)-1H-indazole

Supplier: Apollo Scientific
Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-6-amine, 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

OR43554-1G OR43554-500MG
APOSOR43554-1GEA 302 EUR
APOSOR43554-1G APOSOR43554-500MG
6-Amino-1-(tetrahydropyranyl)-1H-indazole
6-Amino-1-(tetrahydropyranyl)-1H-indazole
Formula: C₁₂H₁₅N₃O
MW: 217,27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699171
CAS Number: 1053655-59-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR