6-Bromo-2-methyl-2H-indazole
Supplier: Apollo Scientific
|
Warning
|
OR43559-1G
OR43559-250MG
APOSOR43559-1GEA
29
EUR
APOSOR43559-1G
APOSOR43559-250MG
6-Bromo-2-methyl-2H-indazole
6-Bromo-2-methyl-2H-indazole
Formula:
C₈H₇BrN₂ MW: 211,0600 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09870048 CAS Number: 590417-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...