7-Bromo-2-methyl-2H-indazole
Supplier: Apollo Scientific
|
Warning
|
OR43561-1G
OR43561-500MG
APOSOR43561-1GEA
78.6
EUR
APOSOR43561-1G
APOSOR43561-500MG
7-Bromo-2-methyl-2H-indazole
7-Bromo-2-methyl-2H-indazole
Formula:
C₈H₇BrN₂ MW: 211,600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09870049 CAS Number: 701910-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...