4-Methyl-3,5-dinitrobenzoic acid

Supplier: Apollo Scientific

Synonyms: 3,5-Dinitro-p-toluic Acid, 3,5-Dinitro-4-toluic acid

OR43583-5G OR43583-10G
APOSOR43583-5GEA 58.2 EUR
APOSOR43583-5G APOSOR43583-10G
4-Methyl-3,5-dinitrobenzoic acid
4-Methyl-3,5-dinitrobenzoic acid
Formula: C₈H₆N₂O₆
MW: 226,14 g/mol
Melting Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00007175
CAS Number: 16533-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR