6-Bromobenzo[d]oxazole 98%

Supplier: Apollo Scientific
Warning

OR43655-5G
APOSOR43655-5GEA 150 EUR
APOSOR43655-5G
6-Bromobenzo[d]oxazole 98%
6-Bromobenzo[d]oxazole
Formula: C₇H₄BrNO
MW: 198,0168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03092891
CAS Number: 375369-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR