1-Methyl-1H-indazole-3-carboxylic acid 99%

Supplier: Apollo Scientific
Warning

OR4383-25G OR4383-5G OR4383-100G
APOSOR4383-25GEA 94.5 EUR
APOSOR4383-25G APOSOR4383-5G APOSOR4383-100G
1-Methyl-1H-indazole-3-carboxylic acid 99%
1-Methyl-1H-indazole-3-carboxylic acid
Formula: C₉H₈N₂O₂
MW: 176,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272569
CAS Number: 50890-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR