Imidazo[1,2-a]pyridin-6-ylmethanol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-(Hydroxymethyl)imidazo[1,2-a]pyridine
OR4418-1G
OR4418-250MG
OR4418-5G
APOSOR4418-1GEA
108
EUR
APOSOR4418-1G
APOSOR4418-250MG
APOSOR4418-5G
Imidazo[1,2-a]pyridin-6-ylmethanol 97%
Imidazo[1,2-a]pyridin-6-ylmethanol
Formula:
C₈H₈N₂O MW: 148,1600 g/mol Melting Pt: 104…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368521 CAS Number: 132213-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...