Ethyl-1-methyl-1H-imidazole-2-carboxylate 90%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Methylimidazole-2-carboxylic acid ethyl ester, Ethyl 1-methylimidazole-2-carboxylate, Ethyl 1-methyl-1H-imidazole-2-carboxylate, Ethyl 1-Methylimidazole-2-carboxylate

OR4425-1G OR4425-5G OR4425-10G
APOSOR4425-1GEA 21.9 EUR
APOSOR4425-1G APOSOR4425-5G APOSOR4425-10G
Ethyl-1-methyl-1H-imidazole-2-carboxylate 90%
Ethyl-1-methyl-1H-imidazole-2-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154,17 g/mol
Melting Pt: 44…50 °C
Storage Temperature: Freezer
MDL Number: MFCD02683326
CAS Number: 30148-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR