(1-(4-Bromophenyl)cyclopropyl)methanol 96%
Supplier: Apollo Scientific
|
Warning
|
OR451112-1G
OR451112-250MG
APOSOR451112-1GEA
201
EUR
APOSOR451112-1G
APOSOR451112-250MG
(1-(4-Bromophenyl)cyclopropyl)methanol 96%
(1-(4-Bromophenyl)cyclopropyl)methanol
Formula:
C₁₀H₁₁BrO MW: 227,1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07374454 CAS Number: 98480-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...