(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
Supplier: Apollo Scientific
|
Warning
|
OR451180-25G
OR451180-5G
APOSOR451180-25GEA
118
EUR
APOSOR451180-25G
APOSOR451180-5G
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
Formula:
C₅H₉NO₂ MW: 115,13 g/mol Boiling Pt: 147…149 °C/0,06mmHg Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077791 CAS Number: 66673-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...