(±)-4-Phenyl-2-butanol 98%
Supplier: Apollo Scientific
|
Warning
|
OR451309-100G
OR451309-25G
OR451309-500G
APOSOR451309-100GEA
163
EUR
APOSOR451309-100G
APOSOR451309-25G
APOSOR451309-500G
(±)-4-Phenyl-2-butanol 98%
(±)-4-Phenyl-2-butanol
Formula:
(C₆H₅)CH₂CH₂CH(OH)CH₃ MW: 150,22 g/mol Boiling Pt: 132 °C Density: 0,97 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00044349 CAS Number: 2344-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...