Methylsulphonylacetone 98+%

Supplier: Apollo Scientific
Warning

OR451312-100G OR451312-25G OR451312-5G
APOSOR451312-100GEA 557 EUR
APOSOR451312-100G APOSOR451312-25G APOSOR451312-5G
Methylsulphonylacetone 98+%
Methylsulphonylacetone
Formula: C₄H₈O₃S
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00014745
CAS Number: 5000-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR