4-(Allyloxy)benzaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR451327-100G
OR451327-25G
OR451327-5G
APOSOR451327-100GEA
227
EUR
APOSOR451327-100G
APOSOR451327-25G
APOSOR451327-5G
4-(Allyloxy)benzaldehyde 97%
4-(Allyloxy)benzaldehyde
Formula:
C₁₀H₁₀O₂ MW: 162,19 g/mol Boiling Pt: 150…152 °C Density: 1,058 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014133 CAS Number: 40663-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...