(S)-cis-Verbenol

Supplier: Apollo Scientific
Warning

Synonyms: (1S-(1alpha,2beta,5alpha))-4,6,6-Trimethylbicyclo(3.1.1)hept-3-en-2-ol, (1S,2S)-4,6,6-Trimethylbicyclo[3.1.1]hept-3-en-2-ol, Bicyclo(3.1.1)hept-3-en-2-ol, 4,6,6-trimethyl-, (1S,2S,5S)-

OR45137-25G OR45137-5G
APOSOR45137-25GEA 725 EUR
APOSOR45137-25G APOSOR45137-5G
(S)-cis-Verbenol
(S)-cis-Verbenol
Formula: C₁₀H₁₆O
MW: 152,237 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00065444
CAS Number: 18881-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR