Ethyl-7-hydroxycoumarin-4-carboxylate 95+%
Supplier: Apollo Scientific
|
Warning
|
OR4521-1G
OR4521-5G
OR4521-250MG
APOSOR4521-1GEA
49.3
EUR
APOSOR4521-1G
APOSOR4521-5G
APOSOR4521-250MG
Ethyl-7-hydroxycoumarin-4-carboxylate 95+%
Ethyl-7-hydroxycoumarin-4-carboxylate
Formula:
C₁₂H₁₀O₅ Melting Pt: 149…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00174422 CAS Number: 1084-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...