N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride 97%

Supplier: Apollo Scientific

OR452101-1G OR452101-500MG
APOSOR452101-1GEA 445 EUR
APOSOR452101-1G APOSOR452101-500MG
N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride 97%
N,N-Dimethyl-2-pyrrolidinecarboxamide hydrochloride
Formula: C₇H₁₅ClN₂O
MW: 178,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13562148
CAS Number: 1246172-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR