5-Bromo-1-indanone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one

OR4530-5G OR4530-100G OR4530-25G
APOSOR4530-5GEA 21.9 EUR
APOSOR4530-5G APOSOR4530-100G APOSOR4530-25G
5-Bromo-1-indanone 98%
5-Bromo-1-indanone
Formula: C₉H₇BrO
MW: 211,06 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR