Ethyl picolinate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate
OR4533-100G
OR4533-10G
APOSOR4533-100GEA
66.7
EUR
APOSOR4533-100G
APOSOR4533-10G
Ethyl picolinate
Ethyl picolinate
Formula:
C₈H₉NO₂ Boiling Pt: 240…241 °C Melting Pt: 44256 °C Density: 1,119 g/cm³ (25 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 EINECS: 219-758-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...