2-Iodotoluene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Iodo-2-methylbenzene

OR4542-100G OR4542-1KG OR4542-500G
APOSOR4542-100GEA 125 EUR
APOSOR4542-100G APOSOR4542-1KG APOSOR4542-500G
2-Iodotoluene 98%
2-Iodotoluene
Formula: C₇H₇I
MW: 218,04 g/mol
Boiling Pt: 210…212 °C
Density: 1,713 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001042
CAS Number: 615-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR