1-Boc-isonipecotic acid ethyl ester 95+%

Supplier: Apollo Scientific
Warning

OR4553-100G OR4553-25G OR4553-500G OR4553-1KG
APOSOR4553-100GEA 65.4 EUR
APOSOR4553-100G APOSOR4553-25G APOSOR4553-500G APOSOR4553-1KG
1-Boc-isonipecotic acid ethyl ester 95+%
1-Boc-isonipecotic acid ethyl ester
Formula: C₁₃H₂₃NO₄
MW: 257,33 g/mol
Boiling Pt: 120…135 °C/0,5mmHg
Density: 1,046 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01763998
CAS Number: 142851-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR