2'-Benzyloxyacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: 1-[2-(Benzyloxy)phenyl]ethan-1-one

OR4566-25G OR4566-5G
APOSOR4566-25GEA 599 EUR
APOSOR4566-25G APOSOR4566-5G
2'-Benzyloxyacetophenone
2'-Benzyloxyacetophenone
Formula: C₁₅H₁₄O₂
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00017240
CAS Number: 31165-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR