4-Isopropylcinnamic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4-Isopropylphenyl)acrylic acid

OR4567-100G OR4567-1G OR4567-50G
APOSOR4567-100GEA 253 EUR
APOSOR4567-100G APOSOR4567-1G APOSOR4567-50G
4-Isopropylcinnamic acid 98%
4-Isopropylcinnamic acid
Formula: C₁₂H₁₄O₂
MW: 190,24 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00016544
CAS Number: 3368-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR