3-Phenoxyphenethylamine

Supplier: Apollo Scientific
Warning

OR460019-1G OR460019-250MG
APOSOR460019-1GEA 123 EUR
APOSOR460019-1G APOSOR460019-250MG
3-Phenoxyphenethylamine
3-Phenoxyphenethylamine
Formula: C₁₄H₁₅NO
Boiling Pt: 142 °C
Density: 1.08 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 118468-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR