1,4-Diiodobutane
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Tetramethylene diiodide
OR460062-100G
OR460062-25G
APOSOR460062-100GEA
50.9
EUR
APOSOR460062-100G
APOSOR460062-25G
1,4-Diiodobutane
1,4-Diiodobutane
Formula:
C₄H₈I₂ MW: 309,92 g/mol Boiling Pt: 108…110 °C/10mmHg Melting Pt: 44382 °C Density: 2,35 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001099 CAS Number: 628-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...