Naphthalene-1,4,5,8-tetracarboxylic dianhydride, Technical Grade

Supplier: Apollo Scientific
Warning

Synonyms: NTCDA, Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 1,4,5,8-Naphthalenetetracarboxylic dianhydride

OR460075-100G OR460075-10G OR460075-25G
APOSOR460075-100GEA 186 EUR
APOSOR460075-100G APOSOR460075-10G APOSOR460075-25G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride, Technical Grade
Naphthalene-1,4,5,8-tetracarboxylic dianhydride
Formula: C₁₄H₄O₆
MW: 268,18 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006915
CAS Number: 81-30-1
EINECS: 201-342-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR