(R)-(-)-3-Hydroxytetrahydrofuran
Supplier: Apollo Scientific
|
Warning
|
OR4601-5G
OR4601-25G
OR4601-100G
APOSOR4601-5GEA
49.5
EUR
APOSOR4601-5G
APOSOR4601-25G
APOSOR4601-100G
(R)-(-)-3-Hydroxytetrahydrofuran
(R)-(-)-3-Hydroxytetrahydrofuran
Formula:
C₄H₈O₂ Boiling Pt: 181 °C Density: 1,097 g/cm³ (25 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067101 CAS Number: 86087-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...