(S)-(+)-3-Hydroxytetrahydrofuran 99%

Supplier: Apollo Scientific
Warning

OR4603-25G OR4603-5G OR4603-100G
APOSOR4603-25GEA 30.6 EUR
APOSOR4603-25G APOSOR4603-5G APOSOR4603-100G
(S)-(+)-3-Hydroxytetrahydrofuran 99%
(S)-(+)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80 °C/15mmHg
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00064327
CAS Number: 86087-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR