4-Chloropyridine-3-carboxaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Chloronicotinaldehyde
OR46034-1G
OR46034-5G
APOSOR46034-1GEA
23.2
EUR
APOSOR46034-1G
APOSOR46034-5G
4-Chloropyridine-3-carboxaldehyde 97%
4-Chloropyridine-3-carboxaldehyde
Formula:
C₆H₄ClNO MW: 141,56 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04112523 CAS Number: 114077-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...