5-Bromothiazole 97%
Supplier: Apollo Scientific
|
Danger
|
OR46041-1G
OR46041-25G
OR46041-5G
APOSOR46041-1GEA
21.9
EUR
APOSOR46041-1G
APOSOR46041-25G
APOSOR46041-5G
5-Bromothiazole 97%
5-Bromothiazole
Formula:
C₃H₂BrNS MW: 164,03 g/mol Boiling Pt: 80…81 °C/18mmHg Density: 1,835 g/cm³ (25 °C) Flash Pt: 76,7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07787394 CAS Number: 3034-55-7 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...