2,4-Dimethoxybenzylamine 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(2,4-Dimethoxyphenyl)methanamine, (2,4-Dimethoxyphenyl)methanamine
OR46045-100G
OR46045-250G
OR46045-25G
APOSOR46045-100GEA
123
EUR
APOSOR46045-100G
APOSOR46045-250G
APOSOR46045-25G
2,4-Dimethoxybenzylamine 95%
2,4-Dimethoxybenzylamine
Formula:
C₉H₁₃NO₂ MW: 167.21 Boiling Pt: 143 °C/15mmHg Density: 1.113 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052393 CAS Number: 20781-20-8 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...