4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 97%

Supplier: Apollo Scientific
Warning

Synonyms: BOC-L-Aspartic acid 4-benzylester, 4-Benzyl N-Boc-L-aspartate, Boc-Asp(Obzl)-OH, Boc-Asp(OBzl)-OH, N-Boc-L-aspartic Acid 4-Benzyl Ester, N-(tert-Butoxycarbonyl)-L-aspartic Acid 4-Benzyl Ester

OR46046-100G
APOSOR46046-100GEA 65.4 EUR
APOSOR46046-100G
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 97%
4-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Formula: C₁₆H₂₁NO₆
MW: 323,35 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00065564
CAS Number: 7536-58-5
EINECS: 231-406-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR