1,1,2,3-Tetrachloropropene

Supplier: Apollo Scientific
Warning

OR46055-1G OR46055-5G
APOSOR46055-1GEA 90.5 EUR
APOSOR46055-1G APOSOR46055-5G
1,1,2,3-Tetrachloropropene
1,1,2,3-Tetrachloropropene
Formula: C₃H₂Cl₄
MW: 179.86 g/mol
Density: 1.54 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 10436-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR