2-Bromo-4-iodophenol 97%

Supplier: Apollo Scientific
Warning

OR46077-10G OR46077-1G OR46077-5G
APOSOR46077-10GEA 469 EUR
APOSOR46077-10G APOSOR46077-1G APOSOR46077-5G
2-Bromo-4-iodophenol 97%
2-Bromo-4-iodophenol
Formula: C₆H₄BrIO
MW: 298,9038 g/mol
Melting Pt: 48…51 °C
Storage Temperature: Ambient
MDL Number: MFCD11870189
CAS Number: 133430-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR