5-Formyl-2-picoline 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Formyl-6-methylpyridine, 2-Methyl-5-formylpyridine, 6-Methyl-3-pyridinecarboxaldehyde, 6-Methylnicotinaldehyde, 6-Methylpyridine-3-carboxaldehyde
OR46109-1G
OR46109-10G
OR46109-5G
APOSOR46109-1GEA
42.2
EUR
APOSOR46109-1G
APOSOR46109-10G
APOSOR46109-5G
5-Formyl-2-picoline 97%
5-Formyl-2-picoline
Formula:
C₇H₇NO MW: 121.14 g/mol Boiling Pt: 95 °C/20mmHg Density: 1.084 g/cm³ (25 °C) Flash Pt: 87.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD08272279 CAS Number: 53014-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...