5-Methoxypyridin-3-ol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Hydroxy-5-methoxypyridine
OR46115-5G
APOSOR46115-5GEA
85.7
EUR
APOSOR46115-5G
5-Methoxypyridin-3-ol
5-Methoxypyridin-3-ol
Formula:
C6H7NO2 MW: 125,1300 g/mol Melting Pt: 147,5…148.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797672 CAS Number: 109345-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...