2-Bromonicotinonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-3-cyanopyridine

OR46203-25G
APOSOR46203-25GEA 89.9 EUR
APOSOR46203-25G
2-Bromonicotinonitrile 98%
2-Bromonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183,01 g/mol
Melting Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234140
CAS Number: 20577-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR