4-Ethoxybenzyl alcohol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Ethoxyphenyl)methanol
OR46213-10G
OR46213-50G
OR46213-100G
APOSOR46213-10GEA
36.4
EUR
APOSOR46213-10G
APOSOR46213-50G
APOSOR46213-100G
4-Ethoxybenzyl alcohol 98%
4-Ethoxybenzyl alcohol
Formula:
C₉H₁₂O₂ MW: 152,19 g/mol Boiling Pt: 272…274 °C Melting Pt: 32…34 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004655 CAS Number: 6214-44-4 EINECS: 228-283-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...