(3R)-(-)-1-Benzyl-3-aminopyrrolidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

OR4624-1G OR4624-5G OR4624-250MG
APOSOR4624-1GEA 71.3 EUR
APOSOR4624-1G APOSOR4624-5G APOSOR4624-250MG
(3R)-(-)-1-Benzyl-3-aminopyrrolidine 97%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine

Chiral intermediate for pharmaceuticals and agrochemicals.

Formula: C₁₁H₁₆N₂
MW: 176,26 g/mol
Boiling Pt: 101…103 °C/3mmHg
Density: 1,02 g/cm³ (25 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00082638
CAS Number: 114715-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR