2-Chloro-5-methylpyrimidine 98%

Supplier: Apollo Scientific
Warning

OR46244-25G OR46244-5G
APOSOR46244-25GEA 72.5 EUR
APOSOR46244-25G APOSOR46244-5G
2-Chloro-5-methylpyrimidine 98%
2-Chloro-5-methylpyrimidine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 89…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09260903
CAS Number: 22536-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR