2,6-Diethynylpyridine 98%

Supplier: Apollo Scientific
Warning

OR46259-1G OR46259-5G OR46259-10G
APOSOR46259-1GEA 81.2 EUR
APOSOR46259-1G APOSOR46259-5G APOSOR46259-10G
2,6-Diethynylpyridine 98%
2,6-Diethynylpyridine
Formula: C₉H₅N
MW: 127.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09039127
CAS Number: 75867-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR