Dimethyl furan-2,5-dicarboxylate 99%
Supplier: Apollo Scientific
|
Warning
|
OR46503-25G
OR46503-100G
APOSOR46503-25GEA
21.9
EUR
APOSOR46503-25G
APOSOR46503-100G
Dimethyl furan-2,5-dicarboxylate 99%
Dimethyl furan-2,5-dicarboxylate
Formula:
C₈H₈O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00092317 CAS Number: 4282-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...