6-Nitropyridin-2-amine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-6-nitropyridine

OR46518-1G
APOSOR46518-1GEA 78.3 EUR
APOSOR46518-1G
6-Nitropyridin-2-amine 98%
6-Nitropyridin-2-amine
Formula: C₅H₅N₃O₂
MW: 139,1121 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD04114202
CAS Number: 14916-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR