Ethyl-4,6-dichloronicotinate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4,6-dichloronicotinate

OR46521-1G OR46521-5G OR46521-25G
APOSOR46521-1GEA 14.7 EUR
APOSOR46521-1G APOSOR46521-5G APOSOR46521-25G
Ethyl-4,6-dichloronicotinate
Ethyl-4,6-dichloronicotinate
Formula: C₈H₇Cl₂NO₂
MW: 220,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234408
CAS Number: 40296-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR