Malonyl dichloride
Supplier: Apollo Scientific
|
Danger
|
OR46548-10G
OR46548-25G
OR46548-100G
APOSOR46548-10GEA
0
EUR
APOSOR46548-10G
APOSOR46548-25G
APOSOR46548-100G
Malonyl dichloride
Malonyl dichloride
Formula:
C₃H₂Cl₂O₂ Boiling Pt: 53…55 °C/19mmHg Density: 1,45 g/cm³ (25 °C) Flash Pt: 47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000735 CAS Number: 1663-67-8 EINECS: 216-772-9 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...