N-Methoxy-N-methylacetamide
Supplier: Apollo Scientific
|
Warning
|
OR46610-100G
OR46610-25G
OR46610-5G
APOSOR46610-100GEA
132
EUR
APOSOR46610-100G
APOSOR46610-25G
APOSOR46610-5G
N-Methoxy-N-methylacetamide
N-Methoxy-N-methylacetamide
Formula:
C₄H₉NO₂ Boiling Pt: 152 °C Density: 0,97 g/cm³ (25 °C) Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060098 CAS Number: 78191-00-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...