3-Dimethylamino-2,2-dimethylpropionaldehyde

Supplier: Apollo Scientific

OR46639-1G OR46639-250MG
APOSOR46639-1GEA 277 EUR
APOSOR46639-1G APOSOR46639-250MG
3-Dimethylamino-2,2-dimethylpropionaldehyde
3-Dimethylamino-2,2-dimethylpropionaldehyde
Formula: C₇H₁₅NO
MW: 129,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043600
CAS Number: 15451-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR