4,5,6,7-Tetrahydro-2H-isoindole-1-carboxylic acid
Supplier: Apollo Scientific
OR46666-100MG
OR46666-50MG
APOSOR46666-100MGEA
292
EUR
APOSOR46666-100MG
APOSOR46666-50MG
4,5,6,7-Tetrahydro-2H-isoindole-1-carboxylic acid
4,5,6,7-Tetrahydro-2H-isoindole-1-carboxylic acid
Formula:
C₉H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09751572 CAS Number: 204068-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...